N,N-Dimethylisobutyramide ≥98.0% (by GC)

Supplier: TCI America
Warning

D4918-25G D4918-5G
TCD4918-25GEA 84 USD
TCD4918-25G TCD4918-5G
N,N-Dimethylisobutyramide ≥98.0% (by GC)
Formula: C₆H₁₃NO
Boiling Pt: 179°C
Flash Pt: 70°C
MDL Number: MFCD00075378
CAS Number: 21678-37-5

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR