Ethyl hydrogen malonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Mono-Ethyl malonate, 3-Ethoxy-3-oxopropanoic acid

M2700-25G M2700-5G
TCM2700-25GEA 124.48 USD
TCM2700-25G TCM2700-5G
Ethyl hydrogen malonate ≥98.0% (by titrimetric analysis)
Ethyl hydrogen malonate
Formula: C₅H₈O₄
MW: 132.115 g/mol
Boiling Pt: 208°C
Melting Pt: -13°C
Flash Pt: 113°C
MDL Number: MFCD00020490
CAS Number: 1071-46-1
EINECS: 213-992-7

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR