Carnosic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (4aR,10aS)-5,6-Dihydroxy-7-isopropyl-1,1-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthrene-4a-carboxylic acid

C3477-100MG C3477-20MG
TCC3477-100MGEA 316.46 USD
TCC3477-100MG TCC3477-20MG
Carnosic acid ≥97.0% (by HPLC, titration analysis)
Carnosic acid
Formula: C₂₀H₂₈O₄
MW: 332.44 g/mol
Melting Pt: 190°C
Storage Temperature: Freezer
MDL Number: MFCD02259459
CAS Number: 3650-09-7

Order Now

Specification Test Results

Appearance White to Yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D +182 to +195 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR