3-Amino-o-anisic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 3-Amino-o-anisic acid, 3-Amino-2-methoxybenzoic acid

A2938-1G A2938-5G
TCA2938-1GEA 137 USD
TCA2938-1G TCA2938-5G
3-Amino-o-anisic acid ≥98.0% (by titrimetric analysis)
3-Amino-o-anisic acid
Formula: C₈H₉NO₃
MW: 167.16
MDL Number: MFCD09701221
CAS Number: 861306-04-9

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR