1,2,6-Hexanetriol ≥99.0% (by GC)
Supplier: TCI America
H1069-25G
H1069-5G
TCH1069-25GEA
168
USD
TCH1069-25G
TCH1069-5G
1,2,6-Hexanetriol ≥99.0% (by GC)
1,2,6-Hexanetriol
Formula:
C₆H₁₄O₃ Boiling Pt: 178°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002976 CAS Number: 106-69-4 EINECS: 203-424-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...