Theobromine 98%

Supplier: Ambeed
Warning

Synonyms: 3,7-Dimethyl-3,7-dihydro-1H-purine-2,6-dione, 3,7-Dimethylxanthine, 3,7-Dihydro-3,7-dimethyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3,7-dimethylpurine

A374174-5G A374174-25G A374174-100G
77616-646EA 6.18 USD
77616-646 77748-749 77748-748
Theobromine 98%
Theobromine

Caution: for Research Use Only

Formula: C₇H₈N₄O₂
MW: 180.17 g/mol
MDL Number: MFCD00022830
CAS Number: 83-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR