(2-Cyclopropylpyridin-4-yl)boronic acid 97%

Supplier: Ambeed
Warning

A118796-1G A118796-250MG
77614-924EA 763.23 USD
77614-924 77614-926
(2-Cyclopropylpyridin-4-yl)boronic acid 97%
(2-Cyclopropylpyridin-4-yl)boronic acid

Caution: for Research Use Only

Formula: C₈H₁₀BNO₂
MDL Number: MFCD11223181
CAS Number: 1189546-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR