L-Tyrosine disodium salt dihydrate (Non-animal/Non-human origin) ≥99% (by titration)
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
(S-3-(4-Hydroxyphenyl)alanine disodium salt dihydrate, Disodium 2-Amino-3-(4-oxidophenyl) propanoate dihydrate, L-3-(4-Hydroxyphenyl)alanine disodium salt dihydrate
00436-250G
00436-100G
00436-1KG
102615-256EA
218.82
USD
102615-256
102615-258
102631-428
L-Tyrosine disodium salt dihydrate (Non-animal/Non-human origin) ≥99% (by titration)
L(-)-Tyrosine disodium salt
Formula:
C₉H₉NO₃Na₂·2H₂O MW: 261.9 |
MDL Number:
MFCD03840558 CAS Number: 122666-87-9 EINECS: 214-684-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...