HOBO® Split-Core AC Current Sensor
Supplier: ONSET COMPUTER CORPORATION SE
CTV-A CTV-B CTV-C CTV-D CTV-E
470346-436EA 198.22 USD
470346-436 470346-430 470346-440 470346-442 470346-434
HOBO® Split-Core AC Current Sensor
Electrodes Other Electrodes
This split-core sensor records AC amperage.
  • UL listed
  • Sensor power induced from line voltage
  • Self-gripping split-core design for fast installation
  • Available in 5 ranges - 20, 50, 100, 200 and 600 A
With an input of AC current, sine wave, single phase 50 or 60 Hz, load power factor 0.5 to 1.0 lead or lag.
Ordering information: Compatible with HOBO U12, UX120-006M, MX1100, and ZW series data loggers with external input channels.