Seraplas® V Valve Filters and Plungers
Supplier: SARSTEDT INC
53.1030 53.420 53.428 53.677
77587-486CS 163.77 USD
77587-486 77587-488 77587-490 77587-492
Seraplas® V Valve Filters and Plungers
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation
The high quality of blood samples collected with the S-Monovette® blood collection system is due to the gentle aspiration technique. It has been proven to significantly reduce hemolysis rates compared to vacuum systems. As a result, erythrocytes remain intact and blood collection does not have to be repeated as often.
  • Reversible separation of serum or plasma and cellular components after centrifugation
  • Mechanical separation body; no diffusion from cellular components in the supernatant (serum or plasma)
  • Stable potassium levels guaranteed
  • Samples with a valve filter in the S-Monovette® can be processed as usual
  • Stable separation layer during storage, transport or shipment
  • For S-Monovettes with diameters of 11 mm (Seraplas® V11), 13 mm (Seraplas® V13), 15 mm (Seraplas® V15) and
  • 16 mm (Seraplas® V16)
Due to the gentle aspiration technique, the S-Monovette® blood collection system is suitable for all vein conditions, from young to old. It offers optimal sample quality and the best results thanks to the revolutionary blood collection system.
This also includes a well-designed, environmentally friendly packaging concept to reduce waste and a ready-to-use system for our needles. All needles can be used straight out of the packaging and allow users to work quickly - there is no assembly required as the needles are available exclusively with integrated holders.
In addition to a premium blood collection system, we support our customers in their everyday work with a pre-analytical consulting group. A global team of experts is available for any pre-analytical questions, areas needing improvement, or product training.