Anti-phospho-MAP2K1 Rabbit Antibody
Supplier: Sino Biological
110470R0008100 110470R000820
77583-514EA 287.37 USD
77583-514 77583-516
Anti-phospho-MAP2K1 Rabbit Antibody
Antibodies
This antibody was obtained from a rabbit immunized with a synthetic phosphopeptide corresponding to residues around Ser218 and Ser222 of the Human MEK1 (Uniprot: Q02750-1) and Ser222 and Ser226 of the Human MEK2 (Uniprot: P36507). Human MEK1 residue Ser218 and Ser222 historically has been referenced as Ser217 and Ser221.