Anti-FCGR3B Mouse Antibody
Supplier: Sino Biological
11046-MM21-100 11046-MM24-100
77583-510EA 277.07 USD
77583-510 77583-512
Anti-FCGR3B Mouse Antibody
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD16b / FCGR3B (rh CD16b / FCGR3B; Catalog#11046-H08H; O75015.2; Met1-Ser200). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.