Anti-IL10 Mouse Antibody
Supplier: Sino Biological
10947-MM03-20 10947-MM07-20
77583-210EA 82.4 USD
77583-210 77583-212
Anti-IL10 Mouse Antibody
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-10 (rh IL-10; Catalog#10947-HNAE; NP_000563.1; Ser 19-Asn 178). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.