InVivoSIM Anti-PD-1 Human Antibody

Supplier: Bio X Cell
SIM0038-100MG SIM0038-1MG SIM0038-25MG SIM0038-50MG SIM0038-5MG
77580-880EA 7984.56 USD
77580-880 77580-882 77580-884 77580-886 77580-888
InVivoSIM Anti-PD-1 Human Antibody
Antibodies
Tislelizumab Biosimilar.

Type:
Antigen: PD-1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR