InVivoSIM Anti-CD25 Human Antibody

Supplier: Bio X Cell
SIM0035-100MG SIM0035-1MG SIM0035-25MG SIM0035-50MG SIM0035-5MG
77580-850EA 7984.56 USD
77580-850 77580-852 77580-854 77580-856 77580-858
InVivoSIM Anti-CD25 Human Antibody
Antibodies
Daclizumab Biosimilar.

Type:
Antigen: CD25
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR