InVivoSIM Anti-IL-6 Human Antibody

Supplier: Bio X Cell
SIM0030-100MG SIM0030-1MG SIM0030-25MG SIM0030-50MG SIM0030-5MG
77580-800EA 7984.56 USD
77580-800 77580-802 77580-804 77580-806 77580-808
InVivoSIM Anti-IL-6 Human Antibody
Antibodies
Siltuximab Biosimilar.

Type:
Antigen: IL-6
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR