InVivoSIM Anti-C5 Human Antibody

Supplier: Bio X Cell
SIM0011-100MG SIM0011-1MG SIM0011-25MG SIM0011-50MG SIM0011-5MG
77580-610EA 7984.56 USD
77580-610 77580-612 77580-614 77580-616 77580-618
InVivoSIM Anti-C5 Human Antibody
Antibodies
Eculizumab Biosimilar.

Type:
Antigen: C5
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype: IgG4
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR