InVivoMAb Mouse IgG2c Isotype Control Anti-Dengue Virus

Supplier: Bio X Cell
BE0366-100MG BE0366-1MG BE0366-25MG BE0366-50MG BE0366-5MG
77579-200EA 4408.4 USD
77579-200 77579-202 77579-204 77579-206 77579-208
InVivoMAb Mouse IgG2c Isotype Control Anti-Dengue Virus
Antibodies
Type: Control
Antigen: IgG2c
Clonality: Monoclonal
Clone: DV5-1
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Dengue virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR