Accessories for Blood Gas Capillaries
Supplier: SARSTEDT INC
65.935.205 65.935.230
77588-348CS 2636.8 USD
77588-348 77588-350
Accessories for Blood Gas Capillaries
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation
For small sample volumes and optimal instrument compatibility, plastic blood gas capillaries are available in a variety of sizes and volumes.
  • Quick-release caps: Good sealing properties due to flexible TPE material, flat shape for improved grip, range of sizes corresponding to outer capillary diameter
  • Mixing wire: Suitable for all SARSTEDT blood gas capillaries
  • Magnet: Bell-shaped for ergonomic handling
Blood gas capillaries provide reliable measurement results with a special calcium-balanced heparin coating. Break-resistant PET plastic ensures maximum safety during collection. The special surface treatment enables quick filling of the capillary, making sample collection easier and reducing the risk of coagulation.