Mailing Boxes
Supplier: SARSTEDT INC
95.900 95.901 95.902 95.903
77589-986CS 75.19 USD
77589-986 77589-988 77589-990 77589-992
Mailing Boxes
Mailers and Shippers
The transport of diagnostics specimens of substance class UN 3373 is subject to legal regulations and guidelines. SARSTEDT offers sample transport systems and secondary containers for a wide variety of transport requirements.
  • Corresponds to the requirement: P 650, ADR, IATA
  • Mailing box for the reliable transport of samples in accordance with packaging instruction P650/IATA/ADR.
  • 4 different sizes can be used variably for all SARSTEDT mailing containers or cold transport containers
  • Various options with an integrated locking mechanism or enclosed foam foil pouch for optimal fixation or padding of secondary containers
  • Customized information e.g. the mailing address can be printed on request
Practical transport of samples and documents by mail.