InVivoSIM Anti-TSLP Human Antibody

Supplier: Bio X Cell
SIM0036-100MG SIM0036-1MG SIM0036-25MG SIM0036-50MG SIM0036-5MG
77580-860EA 7984.56 USD
77580-860 77580-862 77580-864 77580-866 77580-868
InVivoSIM Anti-TSLP Human Antibody
Antibodies
Tezepelumab biosimilar.

Type:
Antigen: TSLP
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR