InVivoSIM Anti-TIGIT Human Antibody

Supplier: Bio X Cell
SIM0033-100MG SIM0033-1MG SIM0033-25MG SIM0033-50MG SIM0033-5MG
77580-830EA 7984.56 USD
77580-830 77580-832 77580-834 77580-836 77580-838
InVivoSIM Anti-TIGIT Human Antibody
Antibodies
Tiragolumab biosimilar.

Type:
Antigen: TIGIT
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR