InVivoPlus Anti-MHC Class I (HLA-A HLA-B HLA-C) Mouse Antibody [clone: W6/32]

Supplier: Bio X Cell
BP0079-100MG BP0079-25MG BP0079-50MG BP0079-5MG
77579-968EA 5735.04 USD
77579-968 77579-970 77579-972 77579-974
InVivoPlus Anti-MHC Class I (HLA-A HLA-B HLA-C) Mouse Antibody [clone: W6/32]
Antibodies
Type: Primary
Antigen: MHC Class I (HLA-A HLA-B HLA-C)
Clonality:
Clone: W6/32
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR