InVivoMAb Anti-CXCL10 Armenian hamster Monoclonal Antibody [clone: 1F11]

Supplier: Bio X Cell
BE0440-100MG BE0440-1MG BE0440-25MG BE0440-50MG BE0440-5MG
77579-940EA 4408.4 USD
77579-940 77579-942 77579-944 77579-946 77579-948
InVivoMAb Anti-CXCL10 Armenian hamster Monoclonal Antibody [clone: 1F11]
Antibodies
Type: Primary
Antigen: CXCL10
Clonality: Monoclonal
Clone: 1F11
Conjugation:
Epitope:
Host: Armenian hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR