InVivoMAb Anti-InVivoMAb anti-Dengue virus type 2 E protein DIII Mouse Monoclonal Antibody [clone: DV2-96]

Supplier: Bio X Cell
BE0432-100MG BE0432-1MG BE0432-25MG BE0432-50MG BE0432-5MG
77579-860EA 4408.4 USD
77579-860 77579-862 77579-864 77579-866 77579-868
InVivoMAb Anti-InVivoMAb anti-Dengue virus type 2 E protein DIII Mouse Monoclonal Antibody [clone: DV2-96]
Antibodies
Type: Primary
Antigen: InVivoMAb anti-Dengue virus type 2 E protein DIII
Clonality: Monoclonal
Clone: DV2-96
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Dengue
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR