InVivoMAb Anti-InVivoMAb anti-human/mouse GRP78 Mouse Monoclonal Antibody [clone: N88]

Supplier: Bio X Cell
BE0416-100MG BE0416-1MG BE0416-25MG BE0416-50MG BE0416-5MG
77579-700EA 4408.4 USD
77579-700 77579-702 77579-704 77579-706 77579-708
InVivoMAb Anti-InVivoMAb anti-human/mouse GRP78 Mouse Monoclonal Antibody [clone: N88]
Antibodies
Type: Primary
Antigen: InVivoMAb anti-human/mouse GRP78
Clonality: Monoclonal
Clone: N88
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR