InVivoMAb Anti-Mac-2 Rat Monoclonal Antibody [clone: M3/38]

Supplier: Bio X Cell
BE0395-100MG BE0395-1MG BE0395-25MG BE0395-50MG BE0395-5MG
77579-490EA 4408.4 USD
77579-490 77579-492 77579-494 77579-496 77579-498
InVivoMAb Anti-Mac-2 Rat Monoclonal Antibody [clone: M3/38]
Antibodies
Type: Primary
Antigen: Mac-2
Clonality: Monoclonal
Clone: M3/38
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR