InVivoMAb Anti-EphA2 Mouse Monoclonal Antibody [clone: B2D6]

Supplier: Bio X Cell
BE0341-100MG BE0341-1MG BE0341-25MG BE0341-50MG BE0341-5MG
77578-960EA 4408.4 USD
77578-960 77578-962 77578-964 77578-966 77578-968
InVivoMAb Anti-EphA2 Mouse Monoclonal Antibody [clone: B2D6]
Antibodies
Type: Primary
Antigen: EphA2
Clonality: Monoclonal
Clone: B2D6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR