Phthalocyanine for organic electronics, purified by sublimation

Supplier: TCI America

Synonyms: Pigment Blue 16, 29H,31H-Phthalocyanine

P2734-100MG P2734-500MG
TCP2734-100MGEA 133.9 USD
TCP2734-100MG TCP2734-500MG
Phthalocyanine for organic electronics, purified by sublimation
Phthalocyanine
Formula: C₃₂H₁₈N₈
MW: 514.55 g/mol
MDL Number: MFCD00005085
CAS Number: 574-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR