PQ401

Supplier: TCI America

Synonyms: 1-(5-Chloro-2-methoxyphenyl)-3-(2-methylquinolin-4-yl)urea, N-(5-Chloro-2-methoxyphenyl)-N’-(2-methyl-4-quinolinyl)urea, IGF-IR Inhibitor

P2479-10MG P2479-50MG
TCP2479-10MGEA 70.04 USD
TCP2479-10MG TCP2479-50MG
PQ401
PQ401
Formula: C₁₈H₁₆ClN₃O₂
MW: 341.8 g/mol
MDL Number: MFCD00160558
CAS Number: 196868-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR