Bergenin

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2R,3S,4S,4aR,10bS)-3,4,8,10-Tetrahydroxy-2-(hydroxymethyl)-9-methoxy-3,4,4a,10b-tetrahydropyrano[3,2-c]isochromen-6(2H)-one

464940010 464942500
TS464940010EA 252 USD
TS464940010 TS464942500
Bergenin
Bergenin

  • Bioactive

Formula: C₁₄H₁₆O₉
MW: 328.27 g/mol
Melting Pt: 232…240 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00133120
CAS Number: 477-90-7

Order Now

Specification Test Results

Appearance White to almost white powder
Infrared spectrum Conforms
Loss on drying ≤6 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR