Estra-5(10),9(11)-diene-3,17-dione-3-ethylene ketal

Supplier: Thermo Scientific Chemicals
Danger

463770010 463770050
TS463770010EA 56 USD
TS463770010 TS463770050
Estra-5(10),9(11)-diene-3,17-dione-3-ethylene ketal
Estra-5(10),9(11)-diene-3,17-dione-3-ethylene ketal

  • Bioactive

Formula: C₂₀H₂₆O₃
MW: 314.43 g/mol
Melting Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD08062585
CAS Number: 5571-36-8
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 152 to 158 °C
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR