Soda lime, granular, pure indicator, with indicator
Supplier: Thermo Scientific Chemicals
|
Danger
|
388350010
388350500
388352500
TS388350010EA
110
USD
TS388350010
TS388350500
TS388352500
Soda lime, granular, pure indicator, with indicator
Soda lime
With indicator white to violet
Storage Temperature:
Ambient |
MDL Number:
MFCD00134094 CAS Number: 8006-28-8 |
Specification Test Results
Appearance | White to beige to light grey Granules |
Loss on drying | ≤20 % (10 g, 200°C, 2 h) |
CO2 absorption capacity | ≥19.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...