Cellulose, microcrystalline ∼ Ø 90 µm, Extra Pure

Supplier: Thermo Scientific Chemicals

382310010 382310050 382310250 382312500
TS382310010EA 118 USD
TS382310010 TS382310050 TS382310250 TS382312500
Cellulose, microcrystalline ∼ Ø 90 µm, Extra Pure
Cellulose
Formula: (C₁₂H₂₂O₁₁)n
Storage Temperature: Ambient
MDL Number: MFCD00081512
CAS Number: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977

Order Now

Specification Test Results

Appearance White to off-white Crystalline or granular powder
Infrared spectrum Conforms
Loss on drying ≤1.5 %
Heavy metals ≤10 ppm
Particle size ≥70 % (> 32 µm)
Residue after ignition ≤0.05 %
pH 5 to 7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR