Copper 99+%, turnings

Supplier: Thermo Scientific Chemicals

317910010 317912500
TS317910010EA 164 USD
TS317910010 TS317912500
Copper 99+%, turnings
Copper
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

Specification Test Results

Appearance Metallic turnings
Total trace metal impurities ≤1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR