Ruthenium 99.9% (trace metals basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Warning

198100010 198100050 198100250
TS198100010EA 183 USD
TS198100010 TS198100050 TS198100250
Ruthenium 99.9% (trace metals basis), powder -200 mesh
Ruthenium
Formula: Ru
MW: 101.07 g/mol
Boiling Pt: 3900 °C (1013 hPa)
Melting Pt: 2310 °C
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1,III
Merck Index: 12,08452

Order Now

Specification Test Results

Appearance Grey Powder
Total trace metal impurities ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR