Platinum, black 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

195210010 195210050
TS195210010EA 380 USD
TS195210010 TS195210050
Platinum, black 98%
Platinum
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now

Specification Test Results

Appearance Black Powder
Assay ≥97.5 % (Pt)
Specific surface area ≥25 m²/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR