Polypropylene glycol, M.W. ∼ 425
Supplier: Thermo Scientific Chemicals
Synonyms:
Poly(propylene oxide), PPG
192150025
192152500
192155000
TS192150025EA
663
USD
TS192150025
TS192152500
TS192155000
Polypropylene glycol, M.W. ∼ 425
Polypropylene glycol
Formula:
H(OCH₂CH₂CH₂)nOH MW: 1025 Boiling Pt: 200 °C (760 mmHg) Melting Pt: –25 °C Density: 1 g/cm³ Flash Pt: 185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00084446 CAS Number: 25322-69-4 |
Specification Test Results
Appearance | Clear viscous liquid |
Infrared spectrum | Conforms |
Water | ≤0.15 % |
Color scale | ≤70 APHA |
Acid value | ≤1 mg KOH/g |
Hydroxyl value | 250 to 276 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...