Poly(N-vinylcarbazole), M.W. ∼90000

Supplier: Thermo Scientific Chemicals

Synonyms: N-Vinylcarbazole Polymer, Poly(9-vinylcarbazole)

179030100 179030250
TS179030100EA 136 USD
TS179030100 TS179030250
Poly(N-vinylcarbazole), M.W. ∼90000
Poly(N-vinylcarbazole)
Density: 1.2 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00134336
CAS Number: 25067-59-8

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR