Polysulfone resin, M.W. ≈. M.W. 60,000, pellets

Supplier: Thermo Scientific Chemicals

178910050 178912500
TS178910050EA 42 USD
TS178910050 TS178912500
Polysulfone resin, M.W. ≈. M.W. 60,000, pellets
Polysulfone resin
Density: 1.24 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00134387
CAS Number: 25135-51-7

Order Now

Specification Test Results

Appearance Amber to beige Pellets
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR