Polyethylene oxide, M.W. 100000-200000

Supplier: Thermo Scientific Chemicals

Synonyms: PEO

178580050 178582500 178585000
TS178580050EA 27 USD
TS178580050 TS178582500 TS178585000
Polyethylene oxide, M.W. 100000-200000
Polyethylene oxide
Formula: (-CH₂CH₂O-)n
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00147970
CAS Number: 25322-68-3

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤2 % (10 g, 50 °C, 5 h, 10 mmHg)
Viscosity 80 to 200 mPa.s (10 % w/w in water at 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR