5-Chlorovaleronitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloropentanenitrile

110140050 110140250
TS110140050EA 29 USD
TS110140050 TS110140250
5-Chlorovaleronitrile 97%
5-Chlorovaleronitrile
Formula: C₅H₈ClN
Boiling Pt: 92…93 °C (10 mmHg)
Density: 1.04 g/cm³
Flash Pt: 98 °C
MDL Number: MFCD00001977
CAS Number: 6280-87-1
EINECS: 228-481-4
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4452 to 1.4472 (20 °C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR