4-Bromobenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

106670250 106671000 106675000
TS106670250EA 92 USD
TS106670250 TS106671000 TS106675000
4-Bromobenzaldehyde 99%
4-Bromobenzaldehyde
Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 244
Melting Pt: 55…58 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00003377
CAS Number: 1122-91-4
EINECS: 214-365-0

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 55 to 59 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR