1-Aminopyrene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyren-1-ylamine, Pyren-1-amine

104530010 104530050
TS104530010EA 85 USD
TS104530010 TS104530050
1-Aminopyrene 97%
1-Aminopyrene
Formula: C₁₆H₁₁N
MW: 217.27 g/mol
Boiling Pt: 415
Melting Pt: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00004140
CAS Number: 1606-67-3
EINECS: 216-521-3

Order Now

Specification Test Results

Appearance Yellow to green Powder
Infrared spectrum Conforms
Melting point 114 to 118 °C
Titration with HClO4 ≥96.0 %
Average molecular weight 70000 (Mn)
Acetyl content 12 to 15 %
Hydroxyl content 1.8 % Typical
Butyryl content 35 to 39 %
Free acid (CH3COOH) ≤0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR