AmberChrom™ chloride form 50WX2 200-400 mesh

Supplier: Thermo Scientific Chemicals

473030025 473031000 473035000
TS473030025EA 1266 USD
TS473030025 TS473031000 TS473035000
AmberChrom™ chloride form 50WX2 200-400 mesh
AmberChrom™ chloride form
Formula: (C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x
CAS Number: 69011-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR