AmberChrom™ hydrogen form 50WX8 200-400 mesh

Supplier: Thermo Scientific Chemicals

472930025 472931000 472935000
TS472930025EA 1398 USD
TS472930025 TS472931000 TS472935000
AmberChrom™ hydrogen form 50WX8 200-400 mesh
AmberChrom™ hydrogen form
MDL Number: MFCD00132722
CAS Number: 69011-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR