AmberChrom™ hydrogen form 50WX8 100-200 mesh

Supplier: Thermo Scientific Chemicals

472920025 472921000 472925000
TS472920025EA 1780 USD
TS472920025 TS472921000 TS472925000
AmberChrom™ hydrogen form 50WX8 100-200 mesh
AmberChrom™ hydrogen form
MDL Number: MFCD00132722
CAS Number: 69011-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR