AmberChrom™ hydrogen form 50WX8 50-100 mesh

Supplier: Thermo Scientific Chemicals

472910025 472911000 472915000
TS472910025EA 1502 USD
TS472910025 TS472911000 TS472915000
AmberChrom™ hydrogen form 50WX8 50-100 mesh
AmberChrom™ hydrogen form
MDL Number: MFCD00132722
CAS Number: 69011-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR