AmberChrom™ hydrogen form 50WX4 200-400 mesh

Supplier: Thermo Scientific Chemicals

472901000 472905000 472900025
TS472901000EA 90 USD
TS472901000 TS472905000 TS472900025
AmberChrom™ hydrogen form 50WX4 200-400 mesh
AmberChrom™ hydrogen form
MDL Number: MFCD00132722
CAS Number: 69011-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR