AmberChrom™ hydrogen form 50WX2 100-200 mesh

Supplier: Thermo Scientific Chemicals

472880025 472881000 472885000
TS472880025EA 1314 USD
TS472880025 TS472881000 TS472885000
AmberChrom™ hydrogen form 50WX2 100-200 mesh
AmberChrom™ hydrogen form
MDL Number: MFCD00132722
CAS Number: 69011-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR